Total Chlorine Checker® Reagents (25 Tests)
Supplier: HANNA INSTRUMENTS CANADA CA
HI711-25 HI711-25
470018-044EA 17.99 CAD
470018-044 CA470018-044
Total Chlorine Checker® Reagents (25 Tests)
Environmental Test Kits
These reagents are the replacement reagents for the total chlorine checker® HC. These high-quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet for traceability.
  • Pre-made reagents for ease of use
  • Prepared with high purity chemicals
  • Marked with expiration date and lot number for traceability
These reagents are high-quality reagents that are pre-measured, allowing for users to achieve fast and accurate measurements with their total chlorine checker® HC. These reagents follow the DPD Method in which the reaction between chlorine and reagent causes a pink tint in the sample. By simply adding the packet of the reagent to the sample, the reaction will take place and the chorine handheld colorimeter will determine the concentration from the color that is produced. The results will be displayed in ppm (mg/L) of total chlorine. These reagents are designed to be used with samples that have an expected range of 0 to 3.50 ppm (mg/L) total chlorine.
Ordering information: For use with chlorine checker (470018-042) only.