Penicillin/Streptomycin/Amphotericin B solution, Corning®
Supplier: Corning
30-004-CI
CA45000-616PK 384.47 CAD
CA45000-616
Penicillin/Streptomycin/Amphotericin B solution, Corning®
Penicillin/Streptomycin/Amphotericin B
This is a combination of penicillin, streptomycin and Amphotericin formulated to contain 10000 units/ml penicillin G, 10 mg/ml streptomycin sulfate and 25 µg/ml amphotericin B.
Penicillin acts by inhibiting bacterial cell-wall synthesis, Streptomycin inhibits prokaryote protein synthesis, and Amphotericin B interferes with fungal membrane permeability.
This combination is effective against Gram-negative and Gram-positive bacteria, fungi and yeasts. The form provided by Mediatech is solubilized in a proprietary citrate buffer and is recommended for use in cell culture applications at 10 ml/L.