Anti-IgG2a Mouse Monoclonal Antibody (Biotin) [clone: MRG2a-83]

Supplier: Biolegend
407503 407504
CA407503-BLEA 132.45 CAD
CA407503-BL CA407504-BL
Anti-IgG2a Mouse Monoclonal Antibody (Biotin) [clone: MRG2a-83]
Antibodies
Biotin anti-rat IgG2a [MRG2a-83]; Isotype: Mouse IgG; Reactivity: Rat; Apps: ELISA, FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG2a
Clonality: Monoclonal
Clone: MRG2a-83
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR