accuro® Gas Detector Pump, Drager
Supplier: Draeger
6400000 4056443
CA34330-068EA 2690.2 CAD
CA34330-068 CA34330-072
accuro® Gas Detector Pump, Drager
Gas Detection Systems
For use with Short-term detector tubes (see 13453-238 series).
  • For use with Short-term detector tubes (see 13453-238 series)
  • Low-cost, one-handed bellows pump is ideal for measurements in hard-to-reach areas
  • Takes short-term measurements with reasonably few strokes when used with Draeger short-term detector tubes
This low-cost, one-handed bellows pump is ideal for measurements in hard-to-reach areas. It takes short-term measurements with reasonably few strokes when used with Draeger short-term detector tubes.
Ordering information: The accuro® gasMore Product Information