E-A-R™ Earmuffs, 3M
Supplier: Avantor
CA33001-588CS 764.41 CAD
CA33001-588 CA89006-770 33001-588
E-A-R™ Earmuffs, 3M
Earmuffs Noise Blocking Earmuffs
3M™ E-A-R™ series earmuffs feature durable, lightweight construction of non-conductive, dielectric plastic that can be worn over the head, behind the neck, or under the chin.
  • Large earcup openings and soft, foam-filled cushions offer a snug, comfortable fit
  • Durable, lightweight construction of non-conductive, dielectric plastic
  • Combine excellent attenuation with comfortable durability
These earmuffs provide a protective seal without excess pressure while the pivoting earcup connections help maintain the seal and proper alignment.