Encore 85 Latex Surgical Gloves, Sterile, Powder-Free, Ansell
Supplier: Ansell Canada
5785000 5785001-1 5785003 5785004-1 5785005 5785006 5785007
CA32918-090CS 1014.42 CAD
CA32918-090 CA32918-091 CA32918-093 CA32918-094 CA32918-095 CA32918-096 CA32918-097
Encore 85 Latex Surgical Gloves, Sterile, Powder-Free, Ansell
Gloves
This natural rubber latex, powder-free surgical glove has a textured finish for superior grip and handling, and its low allergen levels help reduce the risk of latex sensitization in healthcare workers and patients.
  • Low allergen levels
  • Powder-free
  • Textured finish for superior grip
  • Great tactile sensitivity
A textured finish means unequaled reliability when using instruments that require greater gripping capability. The thickness of these gloves is optimized for the best balance of protection and tactile sensitivity, all while allowing for aMore Product Information