Anti-CD267 (TACI) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1A1]
Supplier: Biolegend
311906
CA311906-BLEA 485.12 CAD
CA311906-BL
Anti-CD267 (TACI) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1A1]
Antibodies
PE anti-human CD267 (TACI) [1A1]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.More Product Information