Anti-CD147 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HIM6]
Supplier: Biolegend
306204
CA306204-BLEA 371.82 CAD
CA306204-BL
Anti-CD147 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HIM6]
Antibodies
FITC anti-human CD147 [HIM6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Capuchin Monkey; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.More Product Information