Thermolyne™ Premium Large Muffle Furnaces, Thermo Scientific
Supplier: Thermo Fisher Scientific
F6020C-80 F6028C-80 F6010 F6030CM F6020C F6038CM F6018 F6028C
CA30609-024EA 14758.24 CAD
CA30609-024 CA30609-026 CA30616-051 CA30616-074 CA30616-109 CA30616-154 CA74570-024 CA30616-120
Thermolyne™ Premium Large Muffle Furnaces, Thermo Scientific
Ovens Furnaces (Ashing and Muffle Ovens)
Furnaces for fusions, brazing, enameling, drying precipitates, and volatile solids analysis.
Heating elements are located on four sides for temperature uniformity and are enveloped in custom-formed refractory cement. Reduces the damage caused by contaminant build-up on the resistant wire. Heating elements can be replaced individually, reducing operational cost of replacing all the heating elements. Heating chamber is surrounded by ceramic fiber insulation for maximum energy efficiency.
Economical, easy-to-use digital control with door safety switch and sensor break protection providesMore Product Information