Thermolyne™ Benchtop Muffle Furnaces, Thermo Scientific
Supplier: Thermo Fisher Scientific
F48018 F48058
CA30609-920EA 24880.66 CAD
CA30609-920 CA30610-077 CA30610-191
Thermolyne™ Benchtop Muffle Furnaces, Thermo Scientific
Ovens Furnaces (Ashing and Muffle Ovens)
Furnaces ideal for gravimetric analysis, ashing of organic and inorganic samples, sintering, quantitative analysis and heat treating.
  • Efficient by reducing energy consumption and increase heatup time
  • Reaches a 1200 °C maximum temperature
  • Thermocouple break protection cuts power to heating elements and prevents thermocouple from failure
  • Two open coil heating elements on chamber sides assure fast heat-up with minimum temperature gradient
  • Thermal-efficient ceramic insulation surrounds chamber for maximum energy efficiency
  • F47900 models have 2 L chamber
More Product Information