Anti-CD95 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: DX2]

Supplier: Biolegend
305605 305606
CA305605-BLEA 188.3 CAD
CA305605-BL CA305606-BL
Anti-CD95 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: DX2]
Antibodies
FITC anti-human CD95 (Fas) [DX2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Common Marmoset, Cotton-topped Tamarin, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It isMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD95
Environmentally Preferable
Antigen synonyms FAS|APO-1|TNFRSF6|Fas
Antibody type Primary
Clonality Monoclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Clone DX2
Reactivity Baboon,Capuchin monkey,African green monkey,Cynomolgus monkey,Sooty mangabey,Common marmoset,Rhesus monkey,Cotton-topped tamarin,Pigtailed macaque,Chimpanzee,Human
Host Mouse
Gene ID 355
Isotype IgG1 kappa
Flow cytometry Yes
Immunogen CD95 transfected L cells
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Purification The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR