Anti-CD16 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 3G8]
Supplier: Biolegend
302037 302038
CA302037-BLEA 303.2 CAD
CA302037-BL CA302038-BL
Anti-CD16 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 3G8]
Antibodies
Brilliant Violet 421™ anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Marmoset, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
More Product Information