Minimate™ Tangential Flow Filtration Capsules, Cytiva (Formerly Pall Lab)

Supplier: Cytiva
OA001C12 OA003C12 OA005C12 OA030C12 OA050C12 OA070C12 OA100C12 OA300C12 OA500C12 OA990C12 OAD65C12 OA010C12
CA29301-900EA 451.08 CAD
CA29301-900 CA29301-902 CA29301-904 CA29301-910 CA29301-912 CA29301-914 CA29301-916 CA29301-920 CA29301-922 CA29301-924 CA29301-926 CA29301-908
Minimate™ Tangential Flow Filtration Capsules, Cytiva (Formerly Pall Lab)
Filters Tangential Flow Cassettes
The disposable Minimate™ TFF capsules, with Omega™ membrane, can perform concentration processes desalting or buffer exchange on the same system with minimal user intervention, and achieves high concentration factors in a single step due to the low hold-up volume of the devices.

  • Scaleable
  • Works with a variety of positive pressure laboratory pumps
  • High product recovery
  • 100% integrity tested

The capsules will process samples of up to 1 litre, or efficiently concentrate samples as little as 5 ml.

Delivery information: Supplied with all fittings andMore Product Information
Order Now

SPECIFICATIONS

Dimensions 20 L×3.8 W×1.8 H cm (8×11/2×23/32")
Environmentally Preferable
Effective filtration area 50 cm2
Max. operating pressure 4 bar (60 psi)
Operating temperature range 5...50° C (41...122 °F)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR