Whatman™ Grade 4 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
1004-042 1004-055 1004-070 1004-090 1004-110 1004-125 1004-150 1004-185 1004-240 1004-320 1004-270 1004-400 1004-125 1004-047 1004-047 1004-050 1004-041 1004-917 1004-027 1004-930 1004-041 1004-050 1004-917 1004-930 1004-027 1004-648 1004-648 1004-042 1004-055 1004-070 1004-090 1004-110 1004-150 1004-185 1004-240 1004-320 1004-270 1004-400
28460-019PK 22.99 CAD
28460-019 28460-030 28460-041 28460-063 28460-085 28460-110 28460-120 28460-142 28460-164 28460-171 70240-024 70240-026 CA28460-110 CA11008-514 11008-514 CA11012-602 CA11012-606 CA11012-608 CA11012-828 CA11012-830 10035-226 10035-228 10035-230 10035-232 10035-824 10147-900 CA10147-900 CA28460-019 CA28460-030 CA28460-041 CA28460-063 CA28460-085 CA28460-120 CA28460-142 CA28460-164 CA28460-171 CA70240-024 CA70240-026
Whatman™ Grade 4 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Papers Qualitative Filter Paper
Whatman Grade 4 Qualitative Filter Papers are a standard grade, high flow rate filter paper. They are useful in qualitative analytical separations when fast flow is required and the collection of fine particles is not critical.
  • Excellent retention: Captures coarse particles and gelatinous precipitates such as ferric hydroxide and aluminum hydroxide
  • Fast filtration: Useful for routine clean-up of biological fluids or organic extracts during analysis
These cellulose filters are used in qualitative analytical techniques to determine and identifyMore Product Information