Whatman™ Papers for Healthcare Applications Grade 470, Whatman products (Cytiva)
Supplier: Cytiva
10318493 10318489 10318493 10318489
28153-030PK 1466.48 CAD
28153-030 28153-032 CA28153-030 CA28153-032
Whatman™ Papers for Healthcare Applications Grade 470, Whatman products (Cytiva)
Filters Filter Papers Qualitative Filter Paper
These filter papers can be used for the absorption of culture media, as a blotting paper, for electrophoresis and amino acid chromatography.
  • Soft surface
  • For gelatinous samples
Order Now
SPECIFICATIONS
Environmentally Preferable
Particle retention 20–30 µm
Porosity Coarse
Flow rate Fast