Whatman™ Protran® BA85 & BA83 NC Western Blotting Membranes, Nitrocellulose, Cytiva
Supplier: Cytiva
10401116 10402578 10401316 10401403 10402506 10402525 10402506 10402578 10401316 10401116 10401403 10402525
CA28151-782PK 367.61 CAD
CA28151-782 CA28155-898 CA89026-704 CA89203-144 CA28155-900 28151-818 28155-900 28155-898 89026-704 28151-782 89203-145 CA28151-818
Whatman™ Protran® BA85 & BA83 NC Western Blotting Membranes, Nitrocellulose, Cytiva
Transfer Membranes
Protran® membranes are manufactured from 100% pure nitrocellulose to ensure the highest binding capacity possible. They are compatible with a variety of detection methods, including isotopic, chemiluminescent, colorimetric and fluorescent. No methanol pre-wetting step is required, making this the membrane of choice for proteins which prefer aqueous environments. Prior to transfer the membrane is simply wetted with water, and then placed in the transfer buffer. No other pre treatment steps are necessary.
  • High binding, low background, without the need for stringent washing conditions
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable