Pallflex® Tissuquartz™ Filters
Supplier: Cytiva
7194 7200 7204 7207 7201 7202 7203 7205 7202 7205D 7200D 7202D 7200D
CA21431-416PK 67.41 CAD
CA21431-416 CA28150-897 CA28150-905 CA29301-690 CA28150-899 CA28150-901 CA28150-903 CA28150-907 28150-901 CA76339-872 76411-292 76411-294 CA76411-292
Pallflex® Tissuquartz™ Filters
Filters Glass Fiber Filters
Pallflex Tissuquartz filters are 100% pure quartz filters (with no binder) for air monitoring applications. They feature high flow rate and filtration efficiency.
  • Pure quartz
  • High flow rate
  • Interference-free results
Obtain highly sensitive and interference-free results due to their reduced residual ion and organics content.
Tissuquartz filters can withstand high operating temperature. This feature makes them durable and ideal for analysis of acidic gas, stack sampling, and other harsh testing environments.
For even lower level of trace organics,More Product Information