Versapor® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
66386 66387 66389 66394 66396 66397 66401 66410 66408 60178 66331 66393 66400 66411 66332
CA28149-612PK 199.65 CAD
CA28149-612 CA28149-634 CA28149-638 CA28149-714 CA28149-716 CA28149-718 CA28149-791 CA28149-809 CA28149-860 CA28150-232 CA87003-816 CA28149-610 CA28149-690 CA28149-770 CA28149-810 CA87003-864
Versapor® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Filters Membrane Filters
Ideal for prefiltration of difficult-to-filter solutions and serum.
  • For prefiltration
  • Excellent retention
  • Acrylic copolymer
Designed to be used in applications requiring excellent retention properties combined with tensile strengths in excess of 200 bar (3000 psi).
Filter membrane is made of acrylic copolymer on a nonwoven support. Strong, flexible, and nonshedding, both dry and wet. Delivers high flow rates at low differential pressures. Can be used as a prefilter to replace glass fiber filters. UV or gamma sterilizable.
Ordering information: More Product Information