MicroFunnel™ Plus Disposable Filter Funnels, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4808 4809 4814 4823 4844 4859 4859 4807ME 4829ME
CA28143-570PK 587 CAD
CA28143-570 CA28143-580 CA28143-582 CA28143-586 CA28143-588 CA82028-612 CA87003-810 10057-814 76360-212 76360-214 CA76360-212 CA76360-214 CA77391-976 CA77391-986
MicroFunnel™ Plus Disposable Filter Funnels, Cytiva (Formerly Pall Lab)
Funnels Membrane Filter Funnels
Ideal for monitoring purified water systems maintained at temperatures up to 90ºC (194ºF).
  • Multifunctional
  • Leakproof lid
  • Squeeze separation
  • Available in two capacities
Units function as sample container and filter funnel in one which eliminates a sample transfer step and protects critical samples from contamination
The unique squeeze separation makes membrane retrieval easy. Choose from 0.45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications (not compatible with hot water applications), 0.2 and 0.45 µm Supor® (More Product Information