VacuCap® and VacuCap® PF Bottle-Top Filters, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4621 4624 4631 4634 TA4622 TA4632 4638 4628 4622 4632
CA28143-316PK 488.61 CAD
CA28143-316 CA28143-317 CA28143-330 CA28143-334 CA28143-338 CA28143-340 CA28139-704 CA28139-706 CA28143-315 CA28143-332
VacuCap® and VacuCap® PF Bottle-Top Filters, Sterile, Cytiva (Formerly Pall Lab)
Filters Bottle Top Filters
For sterilization or clarification of laboratory fluids. Ideal for cell culture and serum-containing media.
  • Ideal for cell culture
  • High flow rate
  • Supor® (hydrophilic polyethersulfone) membrane provides high flow rates
  • Acrylic housing, sterile
Polyester membrane support. Effective filtration area of VacuCap 60 PF filters is 30 cm²; VacuCap 90 PF filters, 60 cm². Filters 28143-340 and 28143-338 come with individual attached tubing for each filter device. All other filters come with one piece of tubing per ten filter devices.
Draws directly from mixMore Product Information