Whatman™ Polycap™ HD Disposable Filter Capsules
Supplier: Cytiva
6703-3610 6703-3650 6703-3611 6703-3621 6703-7510 6703-7550 6703-7511 6703-7521 2714T 2612T 2710 2610T 2712M 2810T 2812T 2614T 2711T 2611T 2713T 2712T 2814T 2813 2813T 2813 2610T 2711T 2712M 2712T 2713T 2714T 2810T 2812T 2813T 2814T 2710 2611T 2612T 2614T 6703-3610 6703-3650 6703-3611 6703-3621 6703-7510 6703-7550 6703-7511 6703-7521
28137-888EA 126.06 CAD
28137-888 28137-890 28137-892 28137-894 28137-896 28137-898 28137-900 28137-902 CA11012-832 CA11012-834 CA11012-844 CA11012-852 CA11012-862 CA11012-868 CA11013-004 CA11013-090 CA11013-092 CA11013-228 CA11013-232 CA11013-244 CA11013-700 CA11013-714 CA11013-724 CA11013-726 CA11013-732 CA11013-740 10034-564 10035-360 10035-380 10035-382 10035-384 10035-386 10035-388 10035-414 10035-418 10035-420 10035-426 10035-768 10035-772 10035-776 10035-826 10035-830 10035-832 10035-834 CA28137-888 CA28137-890 CA28137-892 CA28137-894 CA28137-896 CA28137-898 CA28137-900 CA28137-902
Whatman™ Polycap™ HD Disposable Filter Capsules
Filters Membrane Filter Capsules
Polycap HD (Heavy Duty) capsule filter offers a broad range of particle size retention ratings and excellent filtrate purity with polypropylene filters (PP).
  • Polypropylene filters support use with a broad range of solutions, pH solutions, and temperatures
  • Sturdy construction leads to high flow and high retention capacity
  • Can be autoclaved with steam at 121 °C for 20 minutes
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
Chemical processing applications often require a capsule filter to ensure a finishingMore Product Information