Whatman™ Puradisc Syringe Filters, PTFE, Whatman products (Cytiva)
Supplier: Cytiva
6783-0404 6783-1302 6783-1304 6784-1301 6784-1302 6784-1304 6784-1310 6784-2501 6784-2502 6784-2504 6784-2510 6785-2502 6785-2504 6798-2510 6798-2504 6798-2502 6766-1302 6775-1304 6783-0402 6784-0402 6784-0404 6766-1304 6784-0404 6784-1350 6784-1350 6784-0402 6766-1304 6783-0402 6785-2502 6798-2501 10462300 6798-2501 10462300 6783-0404 6783-1302 6783-1304 6784-1301 6784-1302 6784-1304 6784-1310 6784-2501 6784-2502 6784-2504 6784-2510 6785-2504 6798-2510 6798-2504 6798-2502 6766-1302 6775-1304
28137-490PK 1783.19 CAD
28137-490 28137-494 28137-496 28137-526 28137-528 28137-530 28137-532 28137-924 28137-926 28137-928 28137-930 28137-932 28137-934 28297-714 70240-160 70240-162 80082-954 80085-928 80085-930 CA28137-486 CA28137-594 CA28137-596 CA28297-700 28137-596 CA11008-526 11008-526 28137-594 28297-700 28137-486 CA28137-932 CA11012-580 CA11013-474 10035-506 10035-808 CA28137-490 CA28137-494 CA28137-496 CA28137-526 CA28137-528 CA28137-530 CA28137-532 CA28137-924 CA28137-926 CA28137-928 CA28137-930 CA28137-934 CA28297-714 CA70240-160 CA70240-162 CA80082-954 CA80085-928 CA80085-930
Whatman™ Puradisc Syringe Filters, PTFE, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • Eight different material options
  • Choice of 4 mm, 13 mm, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters cater to a wide variety of sample typesMore Product Information