Histo-Orientator™ Hot Plate, TBS®
Supplier: General Data Healthcare
TO-120
CA27900-124EA 3133.08 CAD
CA27900-124
Histo-Orientator™ Hot Plate, TBS®
Slide Warmers/Dryers
Removes wrinkles and bubbles from sections
Also useful with cryostat to ensure attachment of frozen sections to the slide, preventing loosening of the tissue during agitation in hand staining.