Free and Total Chlorine Test Strips, Hach
Supplier: Hach
2745050
CA27450-50EA 62.02 CAD
CA27450-50
Free and Total Chlorine Test Strips, Hach
Environmental Test Kits
The most widely used disinfectant for drinking water, chlorine is also important for sanitizing swimming pools, cooling towers, other industrial equipment, and in the treatment of municipal wastewater. Its measurement and control are vital for both safety and economic reasons. Chlorine exists in water as free chlorine (hypochlorite) or combined chlorine (chloramines).
With Hach Water Quality Test Strips, technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site
Accurate and easy to use - just dip and read. Great for quick, on-site evaluations of free and total chlorine levels.
No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing.