VWR® Deep Petri Dishes
Supplier: VWR International
CA25389-332
CA25389-332CS 363.37 CAD
CA25389-332
VWR® Deep Petri Dishes
Culture Dishes Petri Dishes
These deep-dish Petri dishes accommodate deeper media fills for longer culturing periods.
  • Allow even distribution of culture media
  • Excellent temperature and chemical resistance
  • Bottom and cover are flat inside and out
Suitable for sample purposes in a wide variety of industries.