Sulfuric acid 0.035 N volumetric solution

Supplier: Hach
Danger

2349732
CA23497-32EA 46.66 CAD
CA23497-32
Sulfuric acid 0.035 N volumetric solution
Sulfuric acid

For alkalinity determination by drop-count titration test kit.

Formula: H₂SO₄
MDL Number: MFCD00064589
CAS Number: 7664-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR