Citric Acid F Reagent Solution, Hach
Supplier: Hach
2254249
CA22542-49EA 109.35 CAD
CA22542-49
Citric Acid F Reagent Solution, Hach
Environmental Test Kits
For determination of ultra-low range silica by the Heteropoly Blue method (Hach Method 8282)
Range: 3-1000 μg/L SiO2.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.