Nitrogen-Nitrate Reagent Set, LR, 25 ml, Cadmium Reduction, Hach
Supplier: Hach
2245100
CA22451-00EA 356.21 CAD
CA22451-00
Nitrogen-Nitrate Reagent Set, LR, 25 ml, Cadmium Reduction, Hach
Environmental Test Kits
For determination of Nitrate Nitrogen by the Cadmium Reduction method
Hach Method 8192. Range: up to 0.50 mg/L NO3-N. Sample Size: 25 mL. Reagent Set includes: NitriVer® 3 Reagent Powder Pillows and NitraVer® 6 Reagent Powder Pillows for 100 tests.