ERGOMAT® Complete Smooth Anti-Fatigue Floor Mats, ERGOMAT
Supplier: Ergomat
SX0203 SX0204 SX0205 SX0206 SX0207 SX0208 SX0209 SX0210 SX0211 SX0212 SX0213 SX0214 SX0215 SX0216 SX0217 SX0218 SX0219 SX0220 SX0221 SX0222 SX0223 SX0224 SX0225 SX0226 SX0227 SX0228 SX0229 SX0230 SX0260 SX0303 SX0304 SX0305 SX0306 SX0307 SX0308 SX0309 SX0310 SX0311 SX0312 SX0313 SX0314 SX0315 SX0316 SX0317 SX0318 SX0319 SX0320 SX0321 SX0322 SX0323 SX0324 SX0325 SX0326 SX0327 SX0328 SX0329 SX0330 SX0360 SX0404 SX0405 SX0406 SX0407 SX0408 SX0409 SX0410 SX0411 SX0412 SX0413 SX0414 SX0415 SX0416 SX0417 SX0418 SX0419 SX0420 SX0421 SX0422 SX0423 SX0424 SX0425 SX0426 SX0427 SX0428 SX0429 SX0430 SX0460
CA22230-828EA 219.31 CAD
CA22230-828 CA22230-830 CA22230-832 CA22230-834 CA22230-836 CA22230-838 CA22230-840 CA22230-842 CA22230-844 CA22230-846 CA22230-848 CA22230-850 CA22230-852 CA22230-854 CA22230-856 CA22230-858 CA22230-860 CA22230-862 CA22230-864 CA22230-866 CA22230-868 CA22230-870 CA22230-872 CA22230-874 CA22230-876 CA22230-878 CA22230-880 CA22230-882 CA22230-884 CA22230-886 CA22230-888 CA22230-890 CA22230-892 CA22230-894 CA22230-896 CA22230-898 CA22230-900 CA22230-902 CA22230-904 CA22230-906 CA22230-908 CA22230-910 CA22230-912 CA22230-914 CA22230-916 CA22230-918 CA22230-920 CA22230-922 CA22230-924 CA22230-926 CA22230-928 CA22230-930 CA22230-932 CA22230-934 CA22230-936 CA22230-938 CA22230-940 CA22230-942 CA22230-944 CA22230-946 CA22230-948 CA22230-950 CA22230-952 CA22230-954 CA22230-956 CA22230-958 CA22230-960 CA22230-962 CA22230-964 CA22230-966 CA22230-968 CA22230-970 CA22230-972 CA22230-974 CA22230-976 CA22230-978 CA22230-980 CA22230-982 CA22230-984 CA22230-986 CA22230-988 CA22230-990 CA22230-992 CA22230-994 CA22230-996 CA22230-998
ERGOMAT® Complete Smooth Anti-Fatigue Floor Mats, ERGOMAT
Floor Mats Traction and Anti-Fatigue Floor Mats
The ERGOMAT Complete Smooth is a fire retardant, open cell, virgin polyurethane mat
  • Fire retardant, open cell, virgin polyurethane
  • Tolerates mild exposures of oil and other liquids
  • Silicone free, latex free, anti-static, UV resistant
  • Certified for use in ISO Class 5 (100) cleanrooms
It is designed to accommodate standing workers as well as cart and trolley traffic. It can also tolerate mild exposures of oil and other liquids. This silicone and latex free mat is anti-static, UV resistant and certified for use in ISO Class 5 (100) cleanrooms.