DPD Free Chlorine Reagent Powder Pillows, 5 ml, Hach
Supplier: Hach
2197846
CA21978-46PK 36.78 CAD
CA21978-46
DPD Free Chlorine Reagent Powder Pillows, 5 ml, Hach
Environmental Test Kits
For determination of free chlorine with Hach's Chlorine Visual Test Kits
Sample Size: 5 mL. Range: up to 3.5 mg/L Cl2. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Ordering information: Pack of 50 Powder Pillows.