Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemical
216-1203
CA216-1203EA 555.01 CAD
CA216-1203
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Donkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR