IsoTherm-System® Sample Cooling System
Supplier: Eppendorf
022510053 022510240 022510258
CA20901-646EA 340.65 CAD
CA20901-646 CA20901-655 CA20901-657
IsoTherm-System® Sample Cooling System
Boxes Cooling/Freezing Boxes and Bags
Cool samples safely and efficiently using the Eppendorf® IsoTherm-System. Choose between IsoPacks for 0 °C and –21 °C and use the IsoRack for centrifuging, transporting, or storing, ensuring constant temperature maintenance throughout your workflow.
  • Efficiently cool samples at a constant temperature and then thaw them gently
  • IsoRack suitable for centrifuging in a plate rotor with IsoRack adapter
  • IsoRack serves as a work rack when filling micro test tubes and for provisioning, sorting, distributing, transporting, storing, or freezing samples in micro test
More Product Information