Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: W3/25]

Supplier: Biolegend
201515 201516
CA201515-BLEA 234.58 CAD
CA201515-BL CA201516-BL
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: W3/25]
Antibodies
PE/Cy7 anti-rat CD4 [W3/25]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: W3/25
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR