LB Agar Plates with 150 µg/mL Carbenicillin, Teknova
Supplier: TEKNOVA CA
L6011
CA200066-622PK 1743.14 CAD
CA200066-622
LB Agar Plates with 150 µg/mL Carbenicillin, Teknova
Media for Microbiology Prepared Media for Microbiology
Vented Qtray bioassay tray.
  • 4 pack
  • Sterile
Contains 1% Tryptone, 0.5% yeast extract, 1.0% sodium chloride, 1.5% agar, 150 µg/mL carbenicillin.
Ambient shipping conditions. Refrigerate to store.