Anti-HSP25 Mouse Monoclonal Antibody [clone: G3.1]
Supplier: Enzo Life Sciences
SPA-800D SPA-800F
CA200062-646EA 415.54 CAD
CA200062-646 CA200062-648
Anti-HSP25 Mouse Monoclonal Antibody [clone: G3.1]
Antibodies
Hsp27 is one of the most common members of the highly conserved and ubiquitously expressed family of small heat shock proteins (sHsp), which also includes alphaB-crystallin. It is characterized by a conserved C-terminal alpha-crystallin domain consisting of two anti-parallel beta-sheets that promote oligomer formation required for its primary chaperone function as inhibitor of irreversible protein aggregation. Hsp27 oligomerization is modulated by post-translational phosphorylation of Hsp27 at three serine residues, Ser15, Ser78, and Ser82, by a variety of protein kinases including MAPKAPK-3,More Product Information