Anti-MAP3K7 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAS-ST009E
CA200062-334EA 518.24 CAD
CA200062-334
Anti-MAP3K7 Rabbit Polyclonal Antibody
Antibodies
TAK1 (TGF-beta-activated kinase-1) is a 65 kDa serine/threonine kinase that is a member of the MAPKKK family. TAK1 is involved in the regulation of transcription by the TGF-beta super family, as its kinase activity is stimulated by TGF-beta and bone morphogenetic protein (BMP).

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus TAK1.

Type: Primary
Antigen: MAP3K7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol MAP3K7
Antigen name Mitogen-activated Protein Kinase Kinase Kinase 7
Environmentally Preferable
Antigen synonyms transforming growth factor-beta-activated kinase 1|mitogen-activated protein kinase kinase kinase 7 |TAK1|TGF1a |TGF-beta activated kinase 1|MAP3K7
Antibody type Primary
Clonality Polyclonal
Reactivity Human
Host Rabbit
Isotype
Western blot Yes
ImmunoChemistry Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR