Anti-ITGAM Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M1/70]

Supplier: Rockland Immunochemical
200-502-N79
CA200-502-N79EA 871.01 CAD
CA200-502-N79
Anti-ITGAM Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M1/70]
Antibodies
Anti-CD11b is useful for Immunoprecipitation, immunochemistry, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR