Anti-LY6C1 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RB6-8C5]

Supplier: Rockland Immunochemical
200-502-L54
CA200-502-L54EA 888.01 CAD
CA200-502-L54
Anti-LY6C1 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RB6-8C5]
Antibodies
Anti-Ly-6G/Ly-6C (Gr-1) is useful for immunochemistry, Immunoprecipitation, Western Blot, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: LY6C1
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR