Stereo Microscopes, K Series, Motic
Supplier: Motic
1100200900291 1100200900281 1101001700672 1101001700801 1101005900092 1101005900082
CA19000-204EA 4043.18 CAD
CA19000-204 CA19000-208 CA19000-210 CA19000-216 CA19000-218 CA19000-222 CA19000-224 CA19000-228 CA19000-230 CA19000-250 CA19000-254 CA19000-268 CA19000-270 CA19000-272 CA19000-274 CA19000-276 CA19000-278 CA19000-280 CA19000-282 CA19000-284 CA19000-286 CA19000-288 CA19000-296 CA19000-298
Stereo Microscopes, K Series, Motic
Microscopes
These stereo microscopes feature a pair of 10x, 23mm super widefield eyepieces and a 45° inclined binocular tube with dioptric compensation on both tubes and interpupillary distance adjustment between 54mm and 76mm
Choose from the K-400 series microscopes with a four-step magnification changer at 6x, 12x, 25x, and 50x ratios; the K-500 series microsopes with a five-step magnification changer at 6.4x, 10x, 16x, 25x, and 40x ratios; or the K-700 series microscopes that feature a zoom range at 1:5.2 ratio designed on self-centering dovetail ring mechanism, total parfocality, and absoluteMore Product Information