HyClone™ PF-CHO LS Cell Culture Media, HyClone products (Cytiva)
Supplier: Cytiva
SH30335.01 SH30333.01 SH30333.02
CA16777-546EA 119.57 CAD
CA16777-546 CA76462-522 CA76462-524 CA76635-872 CA76635-886
HyClone™ PF-CHO LS Cell Culture Media, HyClone products (Cytiva)
Cell Culture Media
HyClone™ PF-CHO LS media is designed to support the DHFR selection/amplification system. It has been successfully tested in a variety of cell culture systems, including T-flasks, spinner flasks, and bioreactors.
  • Designed to support the DHFR selection/amplification system
  • Successfully tested in a variety of cell culture systems, including T-flasks, spinner flasks and bioreactors
  • Enable growth of many CHO cells with minimal adaptation
  • PF-CHO LS liquid medium contains 4 mM L-glutamine and 0,1% poloxamer 188
  • PF-CHO MPS powder medium does not contain L-glutamine or poloxamer 188
HyClone™ PF-CHO LS media is ideally suited for the cost-effective manufacture of recombinant proteins for academic and industrial applications, genomics and proteomics, in vitro diagnostics, drug target screening and validation, and manufacturing of pre-clinical lots.