HyClone™ Cosmic Calf Serum, U.S. Origin, HyClone products (Cytiva)
Supplier: Cytiva
SH30087.03 SH30087.04IR25-40 SH30087.04IR30-45 SH30087.02 SH30087.02HI SH30087.04
16777-244EA 272.33 CAD
16777-244 CA76324-884 CA76324-886 CA76324-930 CA16777-242 CA76234-774 CA16777-244 CA82013-582 CA82007-442 CA76234-776
HyClone™ Cosmic Calf Serum, U.S. Origin, HyClone products (Cytiva)
Sera (Serum)
HyClone™ Cosmic calf serum is fortified with iron and naturally derived components for increased performance with a broad spectrum of cell type. Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.
  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing
All components are sourced in the U.S. and are traceable. Studies have shown Cosmic Calf outperforms iron-supplemented bovine calf serum and FBS in several applications, and performs exceptionally wellMore Product Information