HyClone™ Calf Serum, U.S. Origin, Cytiva

Supplier: Cytiva
3007303IR25-40 SH30073.03HI
CA76324-872EA 128.05 CAD
CA76324-872 CA76534-626 CA16777-204 CA76237-688 CA76534-652 CA16777-206 CA76237-690 CA82007-424 CA76237-694
HyClone™ Calf Serum, U.S. Origin, Cytiva
Sera (Serum)
HyClone Bovine Calf Serum, U.S, can be used as an effective alternative to foetal bovine serum (FBS).

  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing
  • Endotoxin Concentration ≤ 10 EU/mL
  • Storage Temperature is –10 °C

Origin is carefully collected, processed and filtered in U.S. abbatoirs. Ensures excellent culture results without safety concerns.

Certifications: Fully traceable and conforms to all USDA regulations.
Order Now

SPECIFICATIONS

Environmentally Preferable
Origin US
Endotoxin ≤10 EU/ml
Hemoglobin <30 mg/dl
Storage temperature Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR