HyClone™ Classical Liquid Media, Dulbeccos Modified Eagles Medium, High Glucose, HyClone products (Cytiva)
Supplier: Cytiva
SH30081.02 SH30081.FS SH30022.02 SH30022.FS SH30022.LS SH30243.01 SH30243.02 SH30285.02 SH30585.02
CA16777-129EA 36.69 CAD
CA16777-129 CA16777-175 CA16777-176 CA16750-082 CA16777-131 CA16750-072 CA16750-112 CA16750-124 CA16777-200 CA16777-198 CA16750-074 CA16750-118 CA16777-406 CA16777-408 CA16777-410 CA16777-413 CA95053-194
HyClone™ Classical Liquid Media, Dulbeccos Modified Eagles Medium, High Glucose, HyClone products (Cytiva)
Cell Culture Media
HyClone™ DMEM high glucose liquid media supports cell growth in research, upstream process development, and cell culture manufacturing processes.
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Efficacy and homogeneity is tested with HyClone™ sera
DMEM media contain increased concentrations of vitamins, minerals, and amino acids compared to traditional basal media. DMEM medium composition also includes ferric nitrate, making this cell culture medium an option for certain cell lines requiring iron for cell growth. HyClone™ DMEM media preparations are available in high or low glucose formulations, as well as with or without L-glutamine, magnesium, and sodium pyruvate. DMEM formulation options may also include phenol red as a pH indicator. All available DMEM media products contain a sodium bicarbonate buffer with or without HEPES to maintain optimal culture pH.
HyClone™ DMEM raw material components are screened through strict quality control testing for lot-to-lot consistency. DMEM liquid media products are hydrated using purified water and undergo sterile filtration.
DMEM is used for a variety of mammalian cell culture manufacturing processes and research applications. DMEM media formulations have supported tumorigenicity studies, virus replication analyses, and investigations into the influence of cell culture media on gene expression. DMEM medium is commonly used for adherent cells and has been the cell culture medium of choice for hybridoma, fibroblast, neuron, and smooth muscle cell lines as well as many other tissues types. DMEM medium is frequently combined with fetal bovine serum (FBS) as DMEM media composition does not include the growth factors, proteins, hormones, or lipids needed to promote cell growth.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 2...8 °C