Origami™ 2 Competent Cells, Expression Strains, MilliporeSigma
Supplier: MilliporeSigma
71344-3 71344-4
CA16003-972EA 378.86 CAD
CA16003-972 CA16003-974
Origami™ 2 Competent Cells, Expression Strains, MilliporeSigma
Competent Cells
A common method for transformation of plasmids into E
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lac UV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilising pET recombinants encoding target proteins that affect cell growth and viability.
The standard volume per tube is 0,2 ml,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable