Rosetta™ 2 Singles™ Competent Cells, EMD Millipore
Supplier: MilliporeSigma
71401-4 71401-3 71400-3 71400-4
CA16003-934EA 993.21 CAD
CA16003-934 CA16003-936 CA80602-816 CA82601-996
Rosetta™ 2 Singles™ Competent Cells, EMD Millipore
Competent Cells
A common method for transformation of DNA plasmids into E. coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen® prepared competent cells. Novagen® Singles™ Competent Cells are designed for ultimate convenience and reliability in plasmid transformation.
  • Hosts carry a chromosomal copy of the T7 RNA polymerase gene
  • Strains are suitable for production of protein from target genes
  • Target genes are cloned in T7 expression vectors,
More Product Information