Nunc® Sampling and Transport System, Thermo Scientific
Supplier: Thermo Fisher Scientific
363282 364238 364246
CA15707-922CS 1024.08 CAD
CA15707-922 CA15707-926 CA15707-928 CA15707-936
Nunc® Sampling and Transport System, Thermo Scientific
Swabs Transport Swabs
A sampling and transport system designed for safe, secure handling and transport of biological specimens.
The system consists of two vials: an inner Universal Container and an outer Transport Container. The inner universal container is a self-standing clear polystyrene tube with molded graduations to 25ml that slips inside the outer transport container. Universal container’s polyethylene cap is specially designed to prevent droplets from adhering to it, reducing the risk of aerosol formations when cap is removed. The outer transport container provides a second liquid barrier to protectMore Product Information