Non Sterile Disposable Specimen Container, Simport Scientific
Supplier: Simport Scientific
C570-12 C570-12
CA15705-350CS 780.21 CAD
CA15705-350 76299-232
Non Sterile Disposable Specimen Container, Simport Scientific
Sample Containers General Purpose Containers
PP container with a PE screw cap.
  • Molded-in graduations up to 128 ml (4 oz)
  • Wide base design for stable reliable use
Heavy-duty thick wall construction of both container and lid ensures a positive leakproof seal time after time. The drip ring on the container reduces the chances of contamination.
Ordering information: Supplied non sterile and packaged in bags of 100 stacked by 10's. Yellow caps packed separately in bags of 100.