Falcon® 96-Well Cell Culture Plates, Corning
Supplier: Corning
351177 351172 353910 353072 353075 353077 353916 353219 353936 353227 353296 353376 353377 353936
CA15705-064CS 367.92 CAD
CA15705-064 CA15705-066 CA62406-015 CA62406-081 CA62406-117 CA62406-121 CA80086-578 CABD353219 CA21100-006 CA25382-200 CA25382-208 CA11008-408 CA11008-410 21100-006
Falcon® 96-Well Cell Culture Plates, Corning
Microplates Cell Culture Plates
Falcon® 96-well microplates are available in a variety of configurations and surface chemistries to meet a wide range of applications with fluorescence, luminescence, radiometric or colorimetric endpoints.
  • Flat bottom, with lid
  • Black wells with clear bottom
  • Working volume: 25 to 340 µl
  • Convenient, peel-open packaging
  • Available with untreated or tissue culture-treated surface chemistries
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
Corning® Primaria™ tissue culture treatment additionally incorporates nitrogen-containingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable