KIMTECH PURE® M7 Face Veil With Knitted Headbands, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Microflex
62757 62751 62757
CA15628-268CS 171.13 CAD
CA15628-268 CA15628-264 83007-443
KIMTECH PURE® M7 Face Veil With Knitted Headbands, KIMBERLY-CLARK PROFESSIONAL®
Masks Cleanroom Masks
Single-use personal veils are designed to be used as a hygienic alternative to reusable garments.
  • Low-linting apertured polyethylene film, filters and minimizes contamination risks
  • Exceptional breathability
  • Headband versions usable with reusable or disposable garment systems
Recommended for ISO Class 7 or higher cleanroom environments.
A covered nose piece allows veils to conform to the nose and cheekbones, ensuring a secure and comfortable fit. Veils are available with snaps or knitted headband. Veils with snaps will fit into most reusable hoods.
More Product Information