Pharmacy Tray, Covidien
Supplier: Cardinal Health
8881535101
CA15141-230CS 26.47 CAD
CA15141-230
Pharmacy Tray, Covidien
Syringes Syringes, Medical
Polystyrene tray contains sterile, single-use syringes
The syringes feature polypropylene barrels and plunger rods, graduated barrel markings, and latex-free plunger tips. Luer lock tip design.